Imprimer
StrandBrite™ Green RNA Quantifying Reagent 200X DMSO Solution
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17610 17611
AATB17610EA 460 EUR
AATB17610 AATB17611
StrandBrite™ Green RNA Quantifying Reagent 200X DMSO Solution
Tests Tests à l'acide nucléique
Detecting and quantitating small amounts of RNA is extremely important for a wide variety of molecular biology procedures such as measuring yields of in vitro transcribed RNA and measuring RNA concentrations before performing Northern blot analysis, S1 nuclease assays, RNase protection assays, cDNA library preparation, reverse transcription PCR, and differential display PCR.
  • Bright Fluorescence
  • High Sensitivity
  • RNA-Specific Binding
Attention: For research use only.