Imprimer
Screen Quest™ TR-FRET No Wash cAMP Assay Kits
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
36380 36379 36381
AATB36380EA 1330 EUR
AATB36380 AATB36379 AATB36381
Screen Quest™ TR-FRET No Wash cAMP Assay Kits
Tests Tests cellulaires
Screen Quest™ TR-FRET No Wash cAMP Assay Kit provides a convenient assay method for monitoring the activation of adenylyl cyclase in G-protein coupled receptor systems.
  • Extremely sensitive detection
  • Contains all necessary components
  • No wash step required
  • Robust and adaptable protocol
  • Longer fluorescence lifetime
Attention: For research use only.