Imprimer
Cell Meter™ Beta-Arrestin Translocation GPCR Signaling Kits
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
36390 36391
AATB36390EA 1090 EUR
AATB36390 AATB36391
Cell Meter™ Beta-Arrestin Translocation GPCR Signaling Kits
Tests Tests cellulaires
Virtually all G protein coupled receptors (GPCRs) rapidly undergo desensitization by a common pathway upon activation by ligand binding.
  • Improved sensitivity and brighter signal
  • Contains all necessary components
  • Robust and adaptable protocol
  • Multiple applications
Attention: For research use only.