ReadiLink™ Cy3 Nick Translation dsDNA Labelling Kit
Marques : AAT BIOQUESTTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
It is compatible with a wide variety of sample materials, including bacterial artificial chromosome (BAC) DNA, human genomic DNA, purified PCR products, supercoiled and linearised plasmid DNA.
- Provides all necessary reagents
- Adjustable ratios
- Wide sample type compatibility
The resulted Cy3-labeled DNAs can be used in a variety of molecular biology techniques such as fluorescence in situ hybridisation (FISH)..
Cdt: Packaged in plastic vials.
Attention: For research use only.