Imprimer
Storage Containers for Fridge, PP
Fournisseur: SEMADENI
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
10145 10147 10148
SEMA10145EA 0 EUR
SEMA10145 SEMA10147 SEMA10148
Storage Containers for Fridge, PP
Boîtes Boîtes de rangement
These polypropylene fridge jars/containers are 100% water, air and aroma tight with integrated green seal and the CLIC & LOCK closure system.
  • Corrugated base keeps food dry
  • Very robust and thick-walled
  • Food-safe polypropylene
  • Scale applied in the corner shows the fill level
  • Microwave and dishwasher safe
The containers are nestable and can thus be stacked inside each other. This saves space in the cupboard when storing the empty containers.
Temperature resistance: −20 to +100 °C
SEMA10145 and SEMA10147 have green seal and SEMA10148 has blue seal.