Imprimer
Helixyte™ Green dsDNA Quantifying Reagent 200X DMSO Solution
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17597 17598
AATB17597EA 460 EUR
AATB17597 AATB17598
Helixyte™ Green dsDNA Quantifying Reagent 200X DMSO Solution
Tests Tests à l'acide nucléique
Helixyte™ Green is an excellent nucleic acid sensor that exhibits large fluorescence enhancement upon binding to dsDNA.
  • Same Spectral Properties To SYBR® Green
  • High Sensitivity
  • Useful For Nucleic Acid Labeling In Gels
Attention: For research use only.