Imprimer
MagaDye™ 561 ddATP
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17062 17066
AATB17062EA 1410 EUR
AATB17062 AATB17066
MagaDye™ 561 ddATP
Réactifs pour acides nucléiques Réactifs pour le séquençage
Sanger sequencing, also known as the chain termination method, is a technique for DNA sequencing based upon the selective incorporation of chain-terminating dideoxynucleotides (ddNTPs) by DNA polymerase.
  • In-house quality control
  • Useful for sanger sequencing applications
Attention: For research use only.