Imprimer
Native Bovine Bovine Serum Albumin (from Serum), Florfenicol
Fournisseur: AAT Bioquest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AATB50035EA 739 EUR
AATB50035
Native Bovine Bovine Serum Albumin (from Serum), Florfenicol
Protéines et Peptides
Florfenicol-BSA conjugate is used for developing anti-florfenicol antibodies that are typically used in an ELISA assay.
  • High immunogenicity
  • Useful for antibody development
  • ELISA assay building block
Attention: For research use only.