Imprimer
Native Bovine Serum Albumin, Chloramphenicol
Fournisseur: AAT Bioquest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AATB50001EA 739 EUR
AATB50001
Native Bovine Serum Albumin, Chloramphenicol
Protéines et Peptides
Chloramphenicol-BSA conjugate is used for developing anti-chloramphenicol antibodies that are typically used in an ELISA assay.
  • Antibody development
  • High immunogenicity
Attention: For research use only.