Imprimer
ReadiLink™ iFluor® 647 Oligo and ssDNA Labeling Kit
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17484 17485
AATB17484EA 353 EUR
AATB17484 AATB17485
ReadiLink™ iFluor® 647 Oligo and ssDNA Labeling Kit
Systèmes de liaison et de marquage des protéines
The kit is optimised for efficient labelling and contains all the essential reagents required for efficient labelling of ssDNA or oligos.
  • Robust protocol
  • Bright labeling
  • Photostable fluorescence
The resulting iFluor® 647-labelled DNA probes are ideally suited for biological applications, e.g., electrophoretic mobility shift assays (EMSA), Northern and Southern blots, colony or in situ hybridizations.
Cdt: Packaged in plastic vials.
Attention: For research use only.