Imprimer
ReadiLink™ iFluor® 488 Nick Translation dsDNA Labeling Kit
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17400 17401
AATB17400EA 544 EUR
AATB17400 AATB17401
ReadiLink™ iFluor® 488 Nick Translation dsDNA Labeling Kit
Systèmes de liaison et de marquage des protéines
It is compatible with a wide variety of sample materials, including bacterial artificial chromosome (BAC) DNA, human genomic DNA, purified PCR products, supercoiled and linearized plasmid DNA.
  • Provides all necessary reagents
  • Adjustable ratios
  • Wide sample type compatibility
The resulted iFluor® 488-labeled DNAs can be used in a variety of molecular biology techniques such as fluorescence in situ hybridization (FISH).
Cdt: Packaged in plastic vials.
Attention: For research use only.