Imprimer
Portelite™ Fluorimetric RNA Quantitation Kit, Optimized for Cytocite™ and Qubit™ Fluorometers*
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17658 17659
AATB17658EA 143 EUR
AATB17658 AATB17659
Portelite™ Fluorimetric RNA Quantitation Kit, Optimized for Cytocite™ and Qubit™ Fluorometers*
Tests Tests à l'acide nucléique
Detecting and quantitating small amounts of RNA is extremely important for a wide variety of biological applications such as measuring yields of in vitro transcribed RNA and measuring RNA concentrations before performing Northern blot analysis, S1 nuclease assays, RNase protection assays, cDNA library preparation, reverse transcription PCR, and differential display PCR.
  • Highly sensitive RNA detection
  • Robust protocol
  • Multiple applications
Attention: For research use only.