Imprimer
Screen Quest™ Fluo-8 No Wash Calcium Assay Kit
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
36314 36316 36315
AATB36314EA 152 EUR
AATB36314 AATB36316 AATB36315
Screen Quest™ Fluo-8 No Wash Calcium Assay Kit
Tests Tests cellulaires
Calcium flux assays are preferred methods in drug discovery for screening G protein coupled receptors (GPCR).
  • 100-250 times fluorescence increase once bound to calcium
  • Contains all necessary components
  • No wash step required
  • Robust and adaptable protocol
  • Brighter and more photoStable signal
Attention: For research use only.