Imprimer

Anti-IgG Goat Polyclonal Antibody (iFluor® 514)

Fournisseur: AAT BIOQUEST
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

16612 16801
AATB16612EA 120 EUR
AATB16612 AATB16801
Anti-IgG Goat Polyclonal Antibody (iFluor® 514)
Anticorps
AAT Bioquest's iFluor® dyes are optimised for labeling proteins, in particular, antibodies.

  • Bright signal
  • Increased photostability
  • Minimal quenching on proteins

Attention: For research use only.

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: iFluor™ 514
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Rabbit
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR