Imprimer
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate) ≥85%
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
Synonyme(s): SMCC
4501 4502
AATB4501EA 120 EUR
AATB4501 AATB4502
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate) ≥85%
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate)
SMCC is a non-cleavable and membrane permeable crosslinker with a spacer arm of 8.3 Å. It contains an amine-reactive succinimidyl ester (SE) and a sulfhydryl-reactive maleimide group. NHS esters react with primary amines at pH 7 to 9 to form stable amide bonds. Maleimides react with sulfhydryl groups at pH 6.5 to 7.5 to form stable thioether bonds. The SE group of SMCC is reacted with lysine residues on the carrier protein, converting them to reactive maleimides. SMCC is a reagent that is widely used for generating stable maleimide-activated carrier proteins that can spontaneously react with sulfhydryls. Alternatively these relatively stable maleimide-activated intermediates may be lyophilized and stored for later conjugation to a hapten.
Formule: C₁₆H₁₈N₂O₆
Poids moléculaire: 334.33 g/mol
Numéro MDL: MFCD00009634
Numéro CAS: 64987-85-5
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=adc5b82fd6acc629d1c3fce25600b47d89493f4b&vid=ccf932f6b93c8f0682db613444667f519a6b133b&prid=web&clid=SAPDAM