Imprimer

5-Carboxytetramethylrhodamine succinimidyl ester (5-TAMRA SE) ≥95%

Marques : AAT BIOQUEST
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Attention

Synonyme(s): TAMRA SE, 5 isomer, 5-TAMRA, SE, 5-TAMRA-N-succinimidyl ester, 5-Carboxytetramethylrhodamine N-succinimidyl ester

373 374 375
AATB373EA 159 EUR
AATB373 AATB374 AATB375
5-Carboxytetramethylrhodamine succinimidyl ester (5-TAMRA SE) ≥95%
5-Carboxytetramethylrhodamine succinimidyl ester (5-TAMRA SE)

The single TAMRA isomers are increasingly preferred for labeling peptides and nucleotides because they give better resolution in HPLC purification that is often required in the conjugation processes.

Formule: C₂₉H₂₅N₃O₇
Numéro CAS: 150810-68-7
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35723e154ab16997dde8ff3da2f15804bf920f7a&vid=47e37e9e49e40c8ae85c6c304111a5f45c82454d&prid=web&clid=SAPDAM

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR