Imprimer
N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate ≥95%
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
Synonyme(s): DABCYL acid, SE, DABCYL N-Succinimidyl Ester, 4-[4-(Dimethylamino)phenylazo]benzoic Acid N-Succinimidyl Ester, 2,5-Dioxopyrrolidin-1-yl 4-((4-(dimethylamino)phenyl)diazenyl)benzoate
2004 2005
AATB2004EA 537 EUR
AATB2004 AATB2005
N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate ≥95%
N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate
DABCYL, SE is the amino-reactive form of DABCYL, and widely used to prepare a variety of FRET-based probes that contain DABCYL.
Formule: C₁₉H₁₈N₄O₄
Poids moléculaire: 366.38 g/mol
Numéro MDL: MFCD00467415
Numéro CAS: 146998-31-4
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3288045e91764d21e55c94400a22af9c4d9c43f3&vid=18c9f4e9f08082b5d78265a3870e5b3b582b111d&prid=web&clid=SAPDAM