Imprimer
iFluor® 546, Succinimidyl Ester
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1048 71048 71521 71571
AATB1048EA 307 EUR
AATB1048 AATB71048 AATB71521 AATB71571
iFluor® 546, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
Attention: For research use only.