Imprimer
iFluor® 660, Succinimidyl Ester
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1032 71032 71511 71561
AATB1032EA 307 EUR
AATB1032 AATB71032 AATB71511 AATB71561
iFluor® 660, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
Attention: For research use only.