iFluor® 660, Succinimidyl Ester
Marques : AAT BIOQUESTTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
- Bright Fluorescence
- High Photostability
- Low Quenching On Proteins
- pH-Independent Signal
Attention: For research use only.