Imprimer
Erlenmeyers, Duran®
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0.620.250 0.624.200 0.624.002 0.624.300 0.624.003 0.624.005 0.619.010.BR 0.619.025.BR 0.619.005.BR 0.620.025 0.620.050 0.622.250.BR 0.622.500.BR 0.622.100.BR 0.620.100 0.620.005 0.622.001.BR 0.622.002.BR 0.624.001.BR 0.620.025.BR 0.620.250.BR 0.620.100.BR 0.620.050.BR 0.621.050.E 0.621.100.E 0.621.200.E 0.621.250.E 0.621.300.E 0.621.500.E 0.621.001.E 0.621.002.E 0.622.001.E 0.622.002.E 0.624.250.E 0.624.300.E 0.624.500.E 0.624.001.E 0.624.002.E 0.620.100.E 0.620.250.E 0.619.005.E 0.619.010.E 0.619.025.E 0.619.050.E 0.619.100.E 0.619.250.E 0.620.005.E 0.620.010.E 0.620.025.E 0.620.050.E 0.622.050.E 0.622.100.E 0.622.200.E 0.622.250.E 0.622.300.E 0.622.500.E
201-0184EA 17 EUR
201-0184 201-0188 WITG0.624.002 201-0189 201-0190 201-0196 WITG0.619.010.BR WITG0.619.025.BR WITG0.619.005.BR WITG0.620.025 WITG0.620.050 WITG0.622.250.BR WITG0.622.500.BR WITG0.622.100.BR WITG0.620.100 201-0181 WITG0.622.001.BR WITG0.622.002.BR WITG0.624.001.BR WITG0.620.025.BR WITG0.620.250.BR WITG0.620.100.BR WITG0.620.050.BR WITG0.621.050.E WITG0.621.100.E WITG0.621.200.E WITG0.621.250.E WITG0.621.300.E WITG0.621.500.E WITG0.621.001.E WITG0.621.002.E WITG0.622.001.E WITG0.622.002.E WITG0.624.250.E WITG0.624.300.E WITG0.624.500.E WITG0.624.001.E WITG0.624.002.E WITG0.620.100.E WITG0.620.250.E WITG0.619.005.E WITG0.619.010.E WITG0.619.025.E WITG0.619.050.E WITG0.619.100.E WITG0.619.250.E WITG0.620.005.E WITG0.620.010.E WITG0.620.025.E WITG0.620.050.E WITG0.622.050.E WITG0.622.100.E WITG0.622.200.E WITG0.622.250.E WITG0.622.300.E WITG0.622.500.E
Erlenmeyers, Duran®
Ballons Flacons Erlenmeyers
DURAN®, borosilicate glass 3.3 or borosilicate glass 3.3 (Economy type), clear or amber.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion
Volume imprint.