Imprimer
GAP Hazardous Material Work Stations
Fournisseur: asecos
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
40566901 40563901 40564901 40567901 40569901 40570901 40573901 40572901 40582901 40581901 40586901 40587901 41044901
MSGA40566901EA 0 EUR
MSGA40566901 MSGA40563901 MSGA40564901 MSGA40567901 MSGA40569901 MSGA40570901 MSGA40573901 MSGA40572901 MSGA40582901 MSGA40581901 MSGA40586901 MSGA40587901 MSGA41044901
GAP Hazardous Material Work Stations
Hottes et coffrets Hottes chimiques
Hazardous material work station, GAP model with robust construction made of chemically resistant, anodised aluminium profiles.
  • Required occupational exposure limits are maintained
  • Avoidance of dangerous explosive atmosphere
  • Used for testing of the air equipment according to EN 14175 Part 3 (5.4.4)
  • Maximum user safety
  • Transparent side panes made from 5 mm (toughened safety glass)
  • Highly effective capturing of hazardous materials inside the work station through special fresh air curtain technology
  • Prepared for connection to the on-site exhaust air system and for electrical supply (230 V/50 Hz)
  • For mounting on existing working surfaces
Standard equipment consists of:
Melamine resin coated rear wall
Exhaust and fresh air monitoring with visual and audible alarm
Interior lighting aranged anti-glare (LED 4000 K neutral white 500 lux on the work surface)
Transparent side walls
Opaque, fold-away deflector
Potential-free switching contact 4-pole (max.400 V/25 A)
Power cord 3 m