Imprimer
Flash Chromatography Systems, C-8XX PURE
Fournisseur: BUCHI
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
11C80500 11C81000 11C81500 11C83000 11C83500 11C85000
11C80500.EA 21200 EUR
11C80500. 11C81000. 11C81500. 11C83000. 11C83500. 11C85000.
Flash Chromatography Systems, C-8XX PURE
Systèmes de chromatographie Systèmes de flash-chromatographie
PURE chromatography systems for flash applications.
  • Benefit from convenient placement of the cartridge/column holder
  • Gain extra bench space with solvent platform
  • Enjoy clean air using a fume enclosure
  • Feel safe with your system thanks to automatic monitoring and settings
  • Perform flexible sample injection thanks to smart design
  • Minimise user training time thanks to simple software
Perform any flash or prep HPLC application easily and safely with our extremely compact pure chromatography systems. The pure platform is fitting to your needs and environment and offers maximal user and sample safety, while using minimal space.
The smart design of the pure chromatography system is fitting to your needs and environment. Thanks to its small footprint, extra solvent platform and the placement of cartridge/column holders in the front of the system, you can save precious laboratory space. The combination of flash and prep HPLC in one instrument, as well as integrated UV and (optional) ELSD technologies offer you unmatched purification possibilities all from one compact system.
W×D×H: 365×570×680 mm
Power consumption: 350 W
Connection voltage: 100 to 240 V±10%
Frequency: 50 to 60 Hz
Fuse: 4 A
Overvoltage category: II
IP code: IP 20
Gas pressure (max.): 8 bar (except model C-805)
Compressed air: Oil and dust free (except model C-805)
USB ports: 3
LAN port (RJ45): 1
RFID reader (cartridges): Yes
Fraction collector bay: Closed
Lighted fraction collector bay: Yes
Solvent level sensor: Yes
Waste level sensor: Yes
Vapour sensor: Yes