Imprimer
Hybridoma Growth Media ISF-I
Marques : BIOCONCEPT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1-57S97-I 1-57S97-K
BCON1-57S97-IEA 21 EUR
BCON1-57S97-I BCON1-57S97-K
Hybridoma Growth Media ISF-I
Milieux de culture cellulaire
ISF-1 is a serum-free, chemically defined medium for hybridoma cell culture and monoclonal antibody production.
It contains glutamine and does not require further supplementation. To protect cells from shear forces during production, surfactant is included in the medium. ISF-1 is not suitable for cholesterol dependent cell lines (e.g. NSO and its varients), without further supplementation of lipopprotein.
The addition of antibiotics should not be a substitute for proper sterile techniques. Therefore, use of antibiotics is in most cases neither necessary nor advised. However, in those instances where antibiotics are desired, ISF-1 has been shown to be compatible with the most used antibiotics (e.g. gentamycin, puromycin and amphotericin B).