Imprimer
LDPE Sliding Lock Bag
Fournisseur: HK PACK KRAUTSCHEID
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1020080 1020083 1020085 1020084 1020088
HKPK1020080EA 508 EUR
HKPK1020080 HKPK1020083 HKPK1020085 HKPK1020084 HKPK1020088
LDPE Sliding Lock Bag
Sachets Sacs de stockage
Slide lock bag are used for sales packaging with the practical slide closure.
  • Clothing such as B. T-shirts, boxer shorts, socks are safely packed
  • Printed with white LD logo
  • Standard with plastic end clamp
  • Freeze-resistant from 50 µm
  • Heat-resistant up to 60°
  • Acid-resistant
  • Transparent material
  • 100% high pressure polyethylene (LDPE)
  • Custom-made (size, thickness, material, individual print) on request