Imprimer
ESD Safety Shoes, Lace-Up, J-nude 2 JALCATCH SAS
Fournisseur: Jallatte
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
00JNI26/35 00JNI26/36 00JNI26/37 00JNI26/38 00JNI26/39 00JNI26/40 00JNI26/41 00JNI26/42 00JNI26/43 00JNI26/44 00JNI26/45 00JNI26/46 00JNI26/47
JALC00JNI26/35EA 149 EUR
JALC00JNI26/35 JALC00JNI26/36 JALC00JNI26/37 JALC00JNI26/38 JALC00JNI26/39 JALC00JNI26/40 JALC00JNI26/41 JALC00JNI26/42 JALC00JNI26/43 JALC00JNI26/44 JALC00JNI26/45 JALC00JNI26/46 JALC00JNI26/47
ESD Safety Shoes, Lace-Up, J-nude 2 JALCATCH SAS
Chaussures et bottes
Low safety shoe, urban sport type, protecting the electronic devices against electrostatic discharge phenomena. Upper made of smooth suede leather with breathable nylon fabric inserts.
  • New PU by BASF® for more flexibility and lightness
  • Gridded outsole for natural foot movement
  • High slip resistance with the grooves in the front and the back of the sole
  • 100% composite materials: lighter than steel, non-magnetic, non-conductive of heat or cold
  • Heel stiffener to reinforce the heel seat
Synthetic nubuck, inserts and collar in technical textiles. Silkscreened Jallatte applies PU reinforcement to the front of the sole. Breathable upper lining in 3D mesh, with honeycomb structure for improved all-round ventilation of the foot and fastdrying. Padded tongue for good instep protection, combined with two side bellows to prevent any dirt from getting into the boots.
Padded ergonomic collar in compact flexible foam, lined for extra comfort and better ankle and Achilles tendon protection. Lace up on 1 pair of eyelets and 3 pairs of fabric loops. Soft and gel footbed made of BASF dynamic polyurethane with heat sensitive memory foam and acts over the entire surface of the foot, cushions pressure points, improves weight distribution and shock absorption caused by impact with the ground.
Anatomical, breathable and fitted with the Link ESDTM system, an innovative and seamless, multi-contact static electricity dissipater. The option of swapping the hygienic insole provided for a SECOSOL® orthopaedic insole. PREM-Alu tip in aluminium type B resisting an impact of an energy of 200 joules. Minimum height after test ≥4 mm in relation to the Type A toe cap in accordance with Standard 22568-2:2019.
Antiperforation inner sole and insert in FleXtane™ By Jallatte material, antistatic, 100% composite. Sewn directly onto the bootstrap and covering 100% of the foot for all-round protection.
Certifications: EN ISO 20345:2011 S1P SRC (SRA+SRB). DGUV 112-191 certified.