Imprimer
SGE OptChem™ Single Taper Inlet Liners
Fournisseur: Trajan Scientific and Medical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0926201701 09262017 0926201501 09262015
SGEA0926201701EA 48.9 EUR
SGEA0926201701 SGEA09262017 SGEA0926201501 SGEA09262015
SGE OptChem™ Single Taper Inlet Liners
Matériel de chromatographie Insets d'injection pour chromatographie
SGE OptChem™ GC inlet liner provide optimal performance with advanced coating technologies in an easy to use format for GC and GCMS.
  • Easy to choose - colour coded by geometry to simplify your selection
  • Easy to use - ready to install touchless packaging with pre-fitted CRS ONE O-rings
  • OptChem™ deactivation is a unique thick film which limits breakdown, ensuring minimal loss of sample and increased system sensitivity
  • Touchless packaging format reduces the risk of contamination during installation
  • Recyclable packaging - less waste to landfill
  • Color coded green
This Trajan SGE Tapered/Gooseneck GC inlet liner is designed for split/splitless injections, offering robust and durable for longer life, less maintenance downtime and reduced maintenance costs.
The Tapered/Gooseneck liner is recommended for the following sample or injection modes: Trace level analysis, active samples, split/splitless injection types.
Certifications: Quality tested to USEPA 8081: organochlorine pesticides, and USEPA 8270: semivolatile organic compounds to ensure a high level of inertness
Informations de commande: For Agilent Intuvo 9000, Agilent 8890, Agilent 8860, Agilent 7890, Agilent 6890, Agilent 6850, Agilent 5890, Agilent 4890, Bruker/Varian 1177, Lucidity GC-FID, PerkinElmer GC 2400, Clarus 690, Clarus 590, Scion 8500, Scion 8300, Thermo Scientific, Thermo Trace 1600, Thermo 1610, Thermo 1300 & Thermo GC Ultra gas chromatographs