Imprimer
Cross Brace, NORM5/25
Fournisseur: HUPFER METALLWERKE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0103072 0103072-1200
HUPF0103072EA 47.9 EUR
HUPF0103072 HUPF0103072-1200
Cross Brace, NORM5/25
Mobilier Étagères
Cross brace made of 25×1,5 mm flat profile (in every 3rd shelf bay).
  • Requires one in every 3rd section
  • Suitable for all shelf widths and uprights