Imprimer
SGE OptChem™ Double Taper Inlet Liners
Fournisseur: Trajan Scientific and Medical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0926201801 09262018 09262002
SGEA0926201801EA 47.5 EUR
SGEA0926201801 SGEA09262018 SGEA09262002
SGE OptChem™ Double Taper Inlet Liners
Matériel de chromatographie Insets d'injection pour chromatographie
The SGE inlet liner range aims to make it simple for all gas chromatographers to select the correct liner.
  • Easy to choose - colour coded by geometry to simplify your selection
  • Easy to use - contains O-rings so ready to go
  • Bottom taper minimizes contact with metal parts of the inlet and focuses sample onto the head of the column
  • Top taper aids in minimizing sample flashback
  • Color coded yellow
Double taper liners are recommended for the following sample or injection modes: trace level analyses, splitless, low boiling point compounds, active compounds.
Informations de commande: Double taper available for Agilent, Thermo Scientific, and Varian/Bruker 1177 and 1078/1079 injectors.