Imprimer
CRS ONE O-Rings for Inlet Liner Seals
Fournisseur: Trajan Scientific and Medical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0726782 0726783 0726786
SGEA0726782EA 23.4 EUR
SGEA0726782 SGEA0726783 SGEA0726786
CRS ONE O-Rings for Inlet Liner Seals
Seals Sealing Rings
The o-ring prevents gas leaks and maintains a uniform flow path through the inlet, where even small leaks can cause sample loss - affecting accuracy and reproducibility of results.
  • Consistently forms a reliable seal, preventing inlet leaks and sample loss
  • Pre-cleaned and conditioned to minimize potential active sites and system contamination
  • Resistant to high temperatures and chemical exposure, ensuring optimal performance, durability and longevity
  • Compatible with multiple instruments
Applications: GC and GC-MS analysis. Ideal for trace level analysis and applications where high levels of cleanliness and sensitivity are required.
FKM o-rings available for industry standard inlet liner sizes and various injectors. Maximum injector temperature of 300 °C.
Cdt: Available in packs of 10.