Imprimer
Masterflex® Single-Use Filling Needles
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX30719-00EA 307 EUR
MFLX30719-00 MFLX30719-02 MFLX30719-04 MFLX30719-06
Masterflex® Single-Use Filling Needles
Fittings and Connectors
A high-quality engineered solution for every application in the fill-finish workflow.
  • Material compliant to USP Class VI, FDA 21 CFR, ADCF
  • Double bagged in ISO Class 7 cleanroom
  • High purity and contamination-free materials
  • Ready to use; increases productivity
  • Straightness <2 mm/M for consistent uniformity
  • Oil-free and adhesive-free construction
  • Repeatable performance with high-speed and accuracy
Filling speed, precision and accuracy are at the heart of the design process. Masterflex® Single Use Filling Needles are a cost‐effective and unique alternative to conventional re‐usable filling needles which require tedious and expensive sterilization and validation processes. They come in a variety of sizes engineered to work with Masterflex® L/S Peristaltic Pump Tubing ranging from L/S 14 to I/P 73 to facilitate a range of filling volumes. The Masterflex® Single Use Filling Needles are easily integrated into new or existing single‐use filling lines where the fluid path must be gamma irradiated or autoclaved. Products have a final cleaning and inspection in an ISO Class 7 cleanroom. All filling needles are cleaned with 70% IPA and vertically dried under HEPA-filtered air. Each needle is inspected before sealing in polybags inside the ISO cleanroom.