Imprimer
Cryogenic Tapes for Frozen Surfaces
Fournisseur: GA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TFS-13C1-50WH TFS-13C1-50YE TFS-13C1-50GA TFS-13C1-50BL TFS-13C1-50OR TFS-13C1-50RE TFS-13C3-100WH TFS-13C3-100YE TFS-13C3-100GA TFS-13C3-100BL TFS-13C3-100OR TFS-13C3-100RE TFS-19C1-50WH TFS-19C1-50YE TFS-19C1-50GA TFS-19C1-50BL TFS-19C1-50OR TFS-19C1-50RE TFS-19C3-100WH TFS-19C3-100YE TFS-19C3-100GA TFS-19C3-100BL TFS-19C3-100OR TFS-19C3-100RE TFS-25C1-50WH TFS-25C1-50YE TFS-25C1-50GA TFS-25C1-50BL TFS-25C1-50OR TFS-25C1-50RE TFS-25C3-100WH TFS-25C3-100YE TFS-25C3-100GA TFS-25C3-100BL TFS-25C3-100OR TFS-25C3-100RE
GAINTFS-13C1-50WHEA 34.9 EUR
GAINTFS-13C1-50WH GAINTFS-13C1-50YE GAINTFS-13C1-50GA GAINTFS-13C1-50BL GAINTFS-13C1-50OR GAINTFS-13C1-50RE GAINTFS-13C3-100WH GAINTFS-13C3-100YE GAINTFS-13C3-100GA GAINTFS-13C3-100BL GAINTFS-13C3-100OR GAINTFS-13C3-100RE GAINTFS-19C1-50WH GAINTFS-19C1-50YE GAINTFS-19C1-50GA GAINTFS-19C1-50BL GAINTFS-19C1-50OR GAINTFS-19C1-50RE GAINTFS-19C3-100WH GAINTFS-19C3-100YE GAINTFS-19C3-100GA GAINTFS-19C3-100BL GAINTFS-19C3-100OR GAINTFS-19C3-100RE GAINTFS-25C1-50WH GAINTFS-25C1-50YE GAINTFS-25C1-50GA GAINTFS-25C1-50BL GAINTFS-25C1-50OR GAINTFS-25C1-50RE GAINTFS-25C3-100WH GAINTFS-25C3-100YE GAINTFS-25C3-100GA GAINTFS-25C3-100BL GAINTFS-25C3-100OR GAINTFS-25C3-100RE
Cryogenic Tapes for Frozen Surfaces
Rubans adhésifs
These tapes are intended for identifying frozen containers and surfaces under deep freeze conditions. Ideal for long-term cryo preservation of samples in liquid nitrogen.
  • Temperature range: −196 to +100 °C (−321 to 212 °F)
  • Quickly adheres to already frozen containers and boxes; ideal for long-term storage in cryogenic and deep freeze conditions
  • Continuous format allows use of as much or little as necessary
  • Available in a variety of colours for easy and convenient colour coding
Cryogenic tape for the identification of already frozen containers such as cryo boxes, cardboard and plastic freezer boxes, as well as tubes and bottles which are difficult to label under deep freeze conditions (−80 °C/−112 °F). Ideal for long-term cryo-preservation in liquid and vapor phase liquid nitrogen (−196 °C/−321 °F), laboratory freezers, and dry ice transportation. These cryo tapes can be inscribed using permanent cryo markers and are available in an assortment of colours for easy identification and colour-coding.