Imprimer
Rectangle and Circle Permanent Deep Freeze Labels for Thermal Transfer Printers
Fournisseur: GA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FJT-103C1-2WH FJT-103C3-4WH FJT-158C1-2WH FJT-172C1-2WH FJT-172C3-4WH FJT-183C1-2WH FJT-183C3-4WH FJT-228C1-2WH FJT52NOTC1-2WH FJT52NOTC3-4WH FJT-70C1-2WH FJT-70C3-4WH
GAINFJT-103C1-2WHEA 88.8 EUR
GAINFJT-103C1-2WH GAINFJT-103C3-4WH GAINFJT-158C1-2WH GAINFJT-172C1-2WH GAINFJT-172C3-4WH GAINFJT-183C1-2WH GAINFJT-183C3-4WH GAINFJT-228C1-2WH GAINFJT52NOTC1-2WH GAINFJT52NOTC3-4WH GAINFJT-70C1-2WH GAINFJT-70C3-4WH
Rectangle and Circle Permanent Deep Freeze Labels for Thermal Transfer Printers
Étiquettes
These permanent labels are designed for the identification of tubes, vials, boxes, and other plastic or cardboard containers stored in ultra-low temperature laboratory freezers.
  • Temperature range: −80 to +110 °C
  • Ideal for long-term storage in deep-freeze conditions (−80 °C)
  • No cure time; can be stored in freezers immediately after application
  • No additional lamination necessary
  • Labels and printout resist spraying with alcohols and other chemicals
  • Printable with all major thermal-transfer printer brands
  • Available in a large range of sizes, colours, and configurations
FJT barcode labels are perfect for identifying small diameter vials, microtubes, Falcon tubes, cell culture plates, microplates, and polypropylene and cardboard freezer boxes. These freezer grade labels are waterproof and tear-proof and will adhere to containers in ultra-low temperature (−80 °C) lab freezers and dry ice without the need for extra lamination.
Available in multiple configurations and sizes, with a permanent adhesive. They are made of a durable thermoplastic film and will resist spraying with alcohols, bleach, detergents, as well as wiping with surface cleaning wipes.