Imprimer
Circle Permanent Deep Freeze Labels for Thermal Transfer Printers
Fournisseur: GA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FJT-106C1-2WH FJT-106C3-2WH FJT-115C1-1WH FJT-115C3-1WH FJT22NOTC1-2WH FJT22NOTC3-4WH FJT53NOTC1-2WH FJT59NPNOT12WH FJT59NPNOT32WH FJT61NPNOT14WH FJT61NPNOT34WH
GAINFJT-106C1-2WHEA 83.6 EUR
GAINFJT-106C1-2WH GAINFJT-106C3-2WH GAINFJT-115C1-1WH GAINFJT-115C3-1WH GAINFJT22NOTC1-2WH GAINFJT22NOTC3-4WH GAINFJT53NOTC1-2WH GAINFJT59NPNOT12WH GAINFJT59NPNOT32WH GAINFJT61NPNOT14WH GAINFJT61NPNOT34WH
Circle Permanent Deep Freeze Labels for Thermal Transfer Printers
Étiquettes
These permanent labels are designed for the identification of tubes, vials, boxes, and other plastic or cardboard containers stored in ultra-low temperature laboratory freezers.
  • Temperature range: −80 to +110 °C
  • Ideal for long-term storage in deep-freeze conditions (−80 °C)
  • No cure time; can be stored in freezers immediately after application
  • No additional lamination necessary
  • Labels and printout resist spraying with alcohols and other chemicals
  • Printable with all major thermal-transfer printer brands
  • Available in a large range of sizes, colours, and configurations
FJT barcode labels are perfect for identifying small diameter vials, microtubes, Falcon tubes, cell culture plates, microplates, and polypropylene and cardboard freezer boxes. These freezer grade labels are waterproof and tear-proof, and will adhere to containers in ultra-low temperature (−80 °C) lab freezers and dry ice without the need for extra lamination.
Available in multiple configurations and sizes, with a permanent adhesive. They are made of a durable thermoplastic film and will resist spraying with alcohols, bleach, detergents, as well as wiping with surface cleaning wipes.