Imprimer
Autoclave-Resistant Cover-Up Thermal Transfer Labels
Fournisseur: GA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BOA-19C1-1SI BOA-19C3-3SI BOA-28C1-1SI BOA-28C3-2SI
GAINBOA-19C1-1SIEA 43.9 EUR
GAINBOA-19C1-1SI GAINBOA-19C3-3SI GAINBOA-28C1-1SI GAINBOA-28C3-2SI
Autoclave-Resistant Cover-Up Thermal Transfer Labels
Étiquettes
Autoclave-resistant blackout labels, designed for high-temperature as well as dry heat and steam sterilisation applications.
  • Temperature range: −60 to 150 °C
  • Unique opaque design allows the labels to permanently obscure all pre-existing information
  • Withstands sterilization in steam autoclaves and dry heat ovens
  • Compatible with all major thermal transfer barcode printers
  • RR-class resin ribbon recommended
A strong permanent adhesive ensures these cover-up autoclave-resistant labels won’t peel or wrinkle, even at elevated temperatures up to 150 °C. These labels are compatible with a range of materials, including metal (stainless steel), glass (Pyrex), and plastic (polypropylene). They can adhere strongly to both flat and curved surfaces.
The labels have a unique opaque design, allowing them to permanently block out previously printed information, including pre-existing text, serialised numbers, and barcodes. Thermal transfer printable, these blackout autoclave-resistant labels are ideal for bags and containers marked with sensitive or confidential information that must be autoclaved before disposal, such as biohazard and radioactive material.