Imprimer
Versilon™ C-544-A IB Tubings
Fournisseur: Saint Gobain Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AZY02019 AZY02029 AZY02064
VERNAZY02019EA 870 EUR
VERNAZY02019 VERNAZY02029 VERNAZY02064
Versilon™ C-544-A IB Tubings
Tuyaux Transfer Tubing
Versilon™ C-544-A IB flexible, reinforced tubing from Saint-Gobain is designed for demanding applications requiring a tight bend radius where other tubes have collapsed and failed. Specially formulated from tough ether-based polyurethane resins, this tubing also retains much of its flexibility even at temperatures as low as −73 °C (−100 °F).
  • Exceptional abrasion and tear resistance
  • Tough braid reinforcement for elevated working pressures
  • Excellent resistance to oils, greases and fuels
  • Retains flexibility in sub-zero environments
Reinforced for elevated pressure, Versilon™ C-544-A IB tubing can easily handle applications requiring large volume transfer for viscosity fluids, pastes, and slurries. It is ideal for applications such as food and cosmetic processing, abrasive and viscous slurry transfer, lubrication and degreaser dispensing, pellet and powder transfer, pneumatic sensory devices, instrumentation control lines and coolant recovery systems.
Hardness: 85 (Shore A)
Certifications: FDA criteria for food contacts ; FDA 21CFR 177.1680 and 177.2600 ; Meets NSF 61 criteria for potable water contact.