Imprimer
Sani-Tech® 3" Tri-Clamp × Hosebarb Adapters
Fournisseur: Saint Gobain Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
KAD300-08H KAD300-12H
VERNKAD300-08HEA 1990 EUR
VERNKAD300-08H VERNKAD300-12H
Sani-Tech® 3" Tri-Clamp × Hosebarb Adapters
Tuyaux Transfer Tubing
The Sani-Tech® 3" tri-clamp adapters come in various configurations for connecting to different types of fittings. These adapters are available in both NPT and Hosebarb styles, allowing versatile connections in fluid transfer systems.
  • Available in both Polypropylene and PVDF
  • All Sani-tech® properties
  • Very large range of accessories
The Sani-Tech products and accessories from the Saint-Gobain brand are renowned for their quality and reliability in the field of fluid distribution and control systems. Designed with advanced materials and top-notchEn savoir plus