Imprimer
Tygon® SPT-3350 Tubings
Fournisseur: Saint Gobain Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
T330O-303 ABW00007 T3306-23-LOT
VERNT330O-303EA 1140 EUR
VERNT330O-303 VERNABW00007 VERNT3306-23-LOT
Tygon® SPT-3350 Tubings
Tuyaux Transfer Tubing
Tygon® SPT-3350 tubing from Saint-Gobain is ideal for food and beverage dispensing. It is three times smoother than other silicone tubing, which reduces extractable content and the risk of particle entrapment, as well as microscopic build-up during fluid transfer - maintains the integrity of the transported food and beverage media.
  • Ultra-smooth inner bore reduces potential for particle entrapment
  • Minimal extractables help maintain fluid integrity
  • Documented biocompatibility for sensitive applications
  • Excellent fluid flow characteristics
A smootherEn savoir plus