Imprimer
Tygon® A-60-C Tubings
Fournisseur: Saint Gobain Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AZT00003-S AZT00004-S AZT00007-S AZT00008-S AZT00009-S AZT00010-S AZT00012-S AZT00017-S AZT00018-S AZT00019-S AZT00022-S AZT00023-S AZT00027-S AZT00028-S AZT00029-S AZT00038-S AZT00046-S AZT00053-S
VERNAZT00003-SEA 224 EUR
VERNAZT00003-S VERNAZT00004-S VERNAZT00007-S VERNAZT00008-S VERNAZT00009-S VERNAZT00010-S VERNAZT00012-S VERNAZT00017-S VERNAZT00018-S VERNAZT00019-S VERNAZT00022-S VERNAZT00023-S VERNAZT00027-S VERNAZT00028-S VERNAZT00029-S VERNAZT00038-S VERNAZT00046-S VERNAZT00053-S
Tygon® A-60-C Tubings
Tuyaux Transfer Tubing
Tygon® A-60-C tubing from Saint-Gobain is formulated to withstand extreme temperatures from −59 to 135 °C (−75 to 275 °F) and will not crack or deteriorate when used in demanding cleaning chemical dispensing applications. Extremely flexible, it resists kinks and retains its shape while installing quickly and easily.
  • Long flex life in peristaltic pumps
  • Compatible with virtually all common sanitisers and cleaners
  • Ozone and UV light resistant
  • Repeatedly autoclavable
  • Chemically compatible with a wide range of food types and cleaners
Tygon® A-60-C tubing simplifies cleaning and sterilisation and is ideal for use in clean-in-place and steam-in-place cleaning and sterilisation systems. It is compatible with virtually all commercial cleaners and sanitisers and can be repeatedly autoclaved for up to five cycle times without affecting its overall service life. Its excellent flexural fatigue resistance makes it the best choice for use in peristaltic pumps often found in dispensing equipment.
Hardness: 61 (Shore A).