Imprimer
Versilic™ Tubings, Silicone
Fournisseur: Saint Gobain Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
760500 760581 760670 760690 760810 760510 760520
VERN760500EA 399 EUR
VERN760500 228-0373 VERN760670 228-0376 228-0379 VERN760510 VERN760520
Versilic™ Tubings, Silicone
Tuyaux Transfer Tubing
This tubing meets European Pharmacopoeia requirements for silicone elastomer for closures and tubing, USP Class VI biological tests and ISO 10993 biocompatibility (systemic toxicity, irritation, sensitisation, citotoxicity, hemocompatibility and Ames test).
  • Meets FDA and BfR positive lists as well as food grade tests according to EC directives
  • This CE marked tubing is a non-invasive medical device
  • May be used for contact up to 29 days
  • Peroxide-cured silicone
  • Biological neutrality
  • FDA/BfR
  • US and European Pharmacopoeia
  • Autoclavable
Versilic tubing is biologically inert and does not inhibit cell culture, it stays flexible down to −50 °C and maintains its chemical, mechanical and electrical performance up to +200 °C. 
Hardness: 62 (Shore A)
Certifications: USP (Class VI) ; USA (FDA; 21CFR part 177-2600).