Imprimer
Pure-Fit® SIB® Smooth Inner Bore PVDF Connector Systems
Fournisseur: Saint Gobain Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PFC250PVDF PFLS250PVDF PFLS375PVDF PFLS500PVDF PFMS250PVDF PFMS375PVDF PFMS500PVDF PFR250X375PVDF PFT250PVDF PFT375PVDF PFX250PVDF PFX375PVDF PFY250PVDF PFY375PVDF PFR250X500PVDF
VERNPFC250PVDFEA 778 EUR
VERNPFC250PVDF VERNPFLS250PVDF VERNPFLS375PVDF VERNPFLS500PVDF VERNPFMS250PVDF VERNPFMS375PVDF VERNPFMS500PVDF VERNPFR250X375PVDF VERNPFT250PVDF VERNPFT375PVDF VERNPFX250PVDF VERNPFX375PVDF VERNPFY250PVDF VERNPFY375PVDF VERNPFR250X500PVDF
Pure-Fit® SIB® Smooth Inner Bore PVDF Connector Systems
Raccords et connecteurs
Pure-Fit SIB eliminates entrapment and leak points that can occur with traditional assembly systems. The unique internal design eliminates voids or dead space that can lead to turbulence or stagnation of fluid flow creating the potential for bacterial growth.
  • Total smooth inner bore (SIB) transition design
  • Eliminates entrapment and leak points that occur with traditional fitting systems
  • Complete 360° compression along the entire fitting system
  • No excessive voids at the interface or in the fluid path
  • Utilises the BarbLock® retention and seal system
  • Compatible with Saint-Gobain Sani-Tech®, C-Flex® and PharMed® flexible tubing
  • PVDF is autoclavable and gamma stable
  • All materials completely animal-free and BPA-free
  • Manufactured and packaged in a certified clean room
  • Validation guide available
Pure-Fit® SIB® fittings are fully compatible with Pure-Fit and BarbLock® retainers that provide complete 360° compression for leak-free connections. It is ideal for applications such as single-use assemblies, biopharmaceutical manufacturing, aseptic and sterile fill lines, filtration, general laboratory, final fill, product sampling and fluid transfer. Also available in gamma stable PP, please see separate product page.
Hardness: 85 (Shore D)
Certifications: USP Class VI and ISO criteria and have been physically tested to meet the most demanding applications and stringent test protocols.