Imprimer
Storage Cabinet
Fournisseur: Palbam
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
JE0950C JE0950
PALBJE0950CEA 0 EUR
PALBJE0950C PALBJE0950
Storage Cabinet
Mobilier Armoires de stockage
Removable shelves and sloping roof design as standard. Available on either casters or leveler legs.
  • 3/4 height cabinets
  • stainless steel 304 with electro-polished finish
L×W×H: 900×500×1500 mm