Imprimer
3M™ Full Facepiece Reusable Respirator
Fournisseur: 3M
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
7100236989 7100237033
MMMA7100236989EA 298 EUR
MMMA7100236989 MMMA7100237033
3M™ Full Facepiece Reusable Respirator
Protection respiratoire Masques réutilisables Masques complets
3M™ Full Facepiece Reusable Respirator is multi-function full-face mask designed to provide durable and comfortable protection against a wide range of respiratory hazards. Use in conjunction with approved 3M powered air systems or as a negative pressure respirator with the 3M™ DT Series filter range, providing protection against a wide range of industrial hazards.
  • Use as a negative-pressure single-filter mask or as a face piece for a powered air respirator
  • Sweat port helps to provide user comfort during long periods of use
  • Visor provides a good field of view and comes in choice of polycarbonate or high-heat material
  • Side-fitting filter port and clear inner mask for good forwards and downwards vision
  • Filter doesn't interfere with front-mounted speech diaphragm to aid in easy communication
For a durable mask that can serve as a face piece for a powered air respirator or as a negative-pressure single-filter mask, use 3M™ Full Facepiece Reusable Respirator FF-300-L. This mask is compatible with the 3M™ Versaflo™ TR-600E, 3M™ Versaflo™ TR-800E, 3M™ PF-602E+, PF-602E-ASB powered air systems as well as the 3M™ DT Series of filters including particulate, gas and vapour and combination filters. The visor (polycarbonate or optional high heat visor) provides a good field of vision and the front-mounted speech diaphragm helps improve communication. There is also a sweat port in the chin pocket which helps to provide added comfort during long periods of wear.
The five-point rubber head harness helps provide a secure, stable fit, and the side-fitting filter and clear inner mask help ensure good downwards visibility.
Suspension type: Five point strap
Certifications: EN 12942 Class TM3 and EN 136 Class 3.