Imprimer
Safety Shoes, Clogs, Active, 87020
Fournisseur: Abeba
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
87020/37 87020/38 87020/39 87020/40 87020/41 87020/42 87020/43 87020/44 87020/45 87020/46 87020/47 87020/48 87020/36
ABEB87020/37EA 47.3 EUR
ABEB87020/37 ABEB87020/38 ABEB87020/39 ABEB87020/40 ABEB87020/41 ABEB87020/42 ABEB87020/43 ABEB87020/44 ABEB87020/45 ABEB87020/46 ABEB87020/47 ABEB87020/48 ABEB87020/36
Safety Shoes, Clogs, Active, 87020
Chaussures et bottes
Anti-static safety clogs made from grained leather with a non foldable, adjustable heel strap and a fuel resistant PU outsole with high slip resistance and a self-cleaning profile.
  • Increased breathability
  • Increased durability of the upper
  • Easy-clean upper
  • Sole resistance to acids and bases
  • Adjustable instep strap with buckle fastening
  • Leather - tear resistance in requirements not less than 120 N
  • Lining - increased water vapor permeability in the requirements of mininum 2,0 mg
Suitable for use in gastronomy/kitchen, horeca, hospitals, pharmaceuticals.
Colour: White, Toe: Thermo plastic resistant to deformation, light, elastic.
Slip resistance on ceramic, coated with SLS solution and on steel, coated with glycerol - slip resistance on ceramic substrate, covered with SLS solution and on steel substrate, covered with glycerol. Antistatic footwear - antistatic properties. Energy absorption in the heel area - energy absorbtion in the heel area. The sole resistant to diesel fuel - sole resistance to diesel fuel.
Certifications: EN ISO 20347:2012; 19/2022/PPE/1439/B edition 1; OB (Basic requirements met); SRC (Slip resistance on ceramic).