Imprimer
Wash Bottles, PFA, Transparent
Fournisseur: VITLAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
108897 108797
VITL108897EA 240 EUR
VITL108897 VITL108797
Wash Bottles, PFA, Transparent
Flacons Pissettes
PFA bottles have excellent chemical resistance and high thermal stability and can be used for critical applications such as trace analysis.
Screw cap made from ETFE, spray tube from FEP. Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube.