Imprimer
Sterile Cleanroom Face Masks with Tyvek® Side Extensions
Fournisseur: KEYSTONE CLEANROOM PRODUCTS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1461STXLDSM25 1461STXLDSMA25
KACC1461STXLDSM25EA 2170 EUR
KACC1461STXLDSM25 KACC1461STXLDSMA25
Sterile Cleanroom Face Masks with Tyvek® Side Extensions
Masks Cleanroom Masks
Sterile cleanroom face masks feature Tyvek® side extensions to prevent skin exposure. Double elastic headband fastening makes it easier to put on compared to ties. Optional fog-free design prevents goggles from fogging and mask slippage.
  • Tyvek® side extensions provide wider coverage area and an improved fit, while also eliminating exposed skin
  • Optional adhesive nose strip prevents goggles from fogging
  • Innovative design prevents mask slippage, reducing contamination risks and significant material and labour costs associated with re-gowning
  • Prevents the common observations noted by inspectors during audits related to facial skin exposure inside aseptic processing areas
  • Packaged in chevron clean-peel Tyvek® pouches
Aseptic operators are more effective and productive when they can see clearly and are comfortable.
Certifications: Certificates of Sterility and Conformance.
Cdt: 25 facemasks per chevron clean-peel Tyvek® pouch. Each package is labelled with a lot number for traceability and an expiration date. Each Tyvek® pouch is contained in a sealed poly bag.