Imprimer
ISO Safety Signs, Emergency Exit
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
138881 198490
BRDY138881EA 7.6 EUR
BRDY138881 BRDY198490
ISO Safety Signs, Emergency Exit
Panneaux
Exit and egress marking for facility identification.
  • Rectangular shape
  • Horizontal markings
  • White on green finish
These B-7541 laminated polyester signs provide emergency and first aid direction to assist in facility identification.