Imprimer
Wide Mouth Stainless Containers
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A856-100 A856-50 A856-75
SAMPA856-100EA 2990 EUR
SAMPA856-100 SAMPA856-50 SAMPA856-75
Wide Mouth Stainless Containers
Barils, tonneaux, fûts
The stainless steel wide mouth containers are perfect for multi purpose drums.
  • Suitable for manufacturing and storing powders, creams and liquids
  • Made from chemically resistant 316L stainless steel
  • Includes 6" tri ferrule lid system giving an excellent water and air tight seal
  • Mirror polish finish
  • Can be easily cleaned
These wide mouth stainless steel containers are perfect for storing powders, creams and liquids. They are made from high quality chemically resistant 316L stainless steel and are designed in such a way as to promote easy cleaning. The tri ferrule lid system means the containers can provide both an air and water tight seal.