Imprimer
316L Stainless Steel Jugs
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A406-500 A406-1000 A406-2000 A406-3000
SAMPA406-500EA 379 EUR
SAMPA406-500 SAMPA406-1000 SAMPA406-2000 SAMPA406-3000
316L Stainless Steel Jugs
Béchers
GMP jugs in 316L stainless steel for easy pouring.
  • Made from high quality 316L stainless steel
  • GMP design - crevice-free, mirror polish, full welds
  • Includes a pouring lip for easy emptying
  • Graduation marks for visual convenience
These high quality jugs are made from 316L stainless steel and have a number of GMP features to making them the best choice for transferring liquids in laboratory and production areas.