Imprimer
Liquid Dip Samplers
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5175A-100 5175A-50
SAMP5175A-100EA 511 EUR
SAMP5175A-100 SAMP5175A-50
Liquid Dip Samplers
Échantillonneurs Préleveurs d'échantillons
The stainless steel liquid dip sampler is used to collect a wide variety of liquid samples from the surface.
  • Made from high quality and chemically resistant 316L stainless steel
  • Different sizes available
  • Convenient hands-free emptying feature
Once the sample has been collected, the liquid dip sampler can simply be pushed against the container allowing for the liquid to be easily emptied.